Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057193.2 | NP_476541.2 | 1 - 2 | 119 | 90 | |
XM_006242864.2 | XP_006242926.1 | 1 - 2 | 297 | 90 | ||
GenBank mRNA | AJ305049.1 | - | 1 - 2 | 119 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |