Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109523.1 | NP_001102993.1 | 1 - 2 | 141 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |