Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012356.1 | NP_001012356.1 | 9 - 10 | 1271 | 105 | |
GenBank mRNA | BC089880.1 | - | 9 - 10 | 1271 | 7366190 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |