Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289778.1 | NP_001276707.1 | 142 | 140 | ||
GenBank mRNA | AB266744.1 | - | 142 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |