Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773229.1 | XP_008771451.1 | 3 - 4 | 889 | 137 | |
XM_008773230.1 | XP_008771452.1 | 4 - 5 | 528 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |