Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257113.2 | XP_006257175.1 | 4 - 5 | 687 | 123 | |
XM_006257114.1 | XP_006257176.1 | 4 - 5 | 703 | 123 | ||
XM_343797.7 | XP_343798.2 | 3 - 4 | 802 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |