Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100542.1 | NP_001094012.1 | 3 - 4 | 282 | 118 | |
GenBank mRNA | AI029347.1 | - | 3 - 4 | 295 | 118 | |
BC166783.1 | - | 3 - 4 | 264 | 7935021 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |