Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177935.2 | NP_808796.1 | 5 - 6 | 781 | 100 | |
GenBank mRNA | AY216593.1 | - | 5 - 6 | 781 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |