Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012860.2 | NP_036992.2 | 5 - 6 | 717 | 64 | |
GenBank mRNA | BC089770.1 | - | 5 - 6 | 717 | 7368255 | 64 |
X15734.1 | - | 5 - 6 | 624 | 64 | ||
X60822.1 | - | 5 - 6 | 803 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |