Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106074.1 | NP_001099544.1 | 5 - 6 | 981 | 83 | |
XM_006252872.2 | XP_006252934.1 | 5 - 6 | 1024 | 83 | ||
XM_006252874.2 | XP_006252936.1 | 5 - 6 | 1024 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |