Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252950.2 | XP_006253012.1 | 1 - 2 | 100 | 78 | |
XM_006252951.2 | XP_006253013.1 | 1 - 2 | 100 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |