Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127590.1 | NP_001121062.1 | 6 - 7 | 553 | 78 | |
XM_006252950.2 | XP_006253012.1 | 6 - 7 | 634 | 78 | ||
XM_006252951.2 | XP_006253013.1 | 6 - 7 | 634 | 78 | ||
XM_008771155.1 | XP_008769377.1 | 6 - 7 | 1151 | 78 | ||
GenBank mRNA | BC160887.1 | - | 6 - 7 | 553 | 7387602 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |