Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005908.1 | NP_001005908.1 | 6 - 7 | 727 | 107 | |
XM_006252786.2 | XP_006252848.1 | 6 - 7 | 981 | 107 | ||
GenBank mRNA | BC083778.1 | - | 6 - 7 | 727 | 7193683 | 107 |
FQ212260.1 | - | 6 - 7 | 927 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |