Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019313.1 | NP_062186.1 | 2 - 3 | 484 | 69 | |
XM_006252933.2 | XP_006252995.1 | 2 - 3 | 517 | 69 | ||
GenBank mRNA | AF000973.1 | - | 2 - 3 | 484 | 69 | |
U69885.1 | - | 2 - 3 | 250 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |