Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047115.1 | NP_001040580.1 | 10 - 11 | 1310 | 63 | |
XM_006252936.2 | XP_006252998.1 | 10 - 11 | 1386 | 63 | ||
XM_008771147.1 | XP_008769369.1 | 10 - 11 | 1386 | 63 | ||
GenBank mRNA | DQ538521.1 | - | 10 - 11 | 1310 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |