Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169100.1 | NP_001162571.1 | 1 - 2 | 142 | 80 | |
XM_006252855.2 | XP_006252917.1 | 1 - 2 | 1062 | 80 | ||
GenBank mRNA | BC158812.1 | - | 1 - 2 | 118 | 80 | |
FQ217580.1 | - | 1 - 2 | 143 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |