Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007684.1 | NP_001007685.1 | 1 - 2 | 78 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |