Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044239.1 | NP_001037704.1 | 9 - 10 | 1121 | 72 | |
GenBank mRNA | BC107903.1 | - | 9 - 10 | 1121 | 7455976 | 72 |
FQ212761.1 | - | 3 - 4 | 282 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |