Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012568.2 | NP_036700.1 | 1 - 2 | 623 | 105 | |
XM_003752039.3 | XP_003752087.1 | 2 - 3 | 247 | 105 | ||
XM_008773152.1 | XP_008771374.1 | 1 - 2 | 616 | 105 | ||
GenBank mRNA | AJ310837.1 | - | 1 - 2 | 72 | 105 | |
X61159.1 | - | 1 - 2 | 623 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |