Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256889.2 | XP_006256951.1 | 8 - 9 | 1127 | 91 | |
GenBank mRNA | FQ219851.1 | - | 7 - 8 | 692 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |