Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109252.1 | NP_001102722.1 | 1 - 2 | 176 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |