Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199256.2 | NP_954865.1 | 4 - 5 | 332 | 76 | |
GenBank mRNA | BC094530.1 | - | 4 - 5 | 332 | 7320508 | 76 |
M96630.1 | - | 4 - 5 | 323 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |