Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025734.1 | NP_001020905.1 | 7 - 8 | 1228 | 57 | |
XM_006237038.1 | XP_006237100.1 | 8 - 9 | 1695 | 57 | ||
XM_006237040.1 | XP_006237102.1 | 8 - 9 | 1521 | 57 | ||
XM_006237042.1 | XP_006237104.1 | 7 - 8 | 1467 | 57 | ||
GenBank mRNA | BC097414.1 | - | 7 - 8 | 1228 | 7454313 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |