Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191579.1 | NP_001178508.1 | 10 - 11 | 894 | 208 | |
XM_006243083.2 | XP_006243145.1 | 9 - 10 | 914 | 184 | ||
XM_008766269.1 | XP_008764491.1 | 9 - 10 | 952 | 184 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |