Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108151.1 | NP_001101621.1 | 1 - 2 | 124 | 111 | |
XM_006243122.2 | XP_006243184.1 | 2 - 3 | 1090 | 111 | ||
XM_008766272.1 | XP_008764494.1 | 2 - 3 | 3895 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |