Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007662.1 | NP_001007663.1 | 1 - 2 | 128 | 124 | |
GenBank mRNA | BC081979.1 | - | 1 - 2 | 128 | 7128203 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |