Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007662.1 | NP_001007663.1 | 6 - 7 | 1106 | 104 | |
GenBank mRNA | BC081979.1 | - | 6 - 7 | 1106 | 7128203 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |