Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109643.2 | NP_001103113.1 | 2 - 3 | 207 | 75 | |
GenBank mRNA | EV775546.1 | - | 2 - 3 | 243 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |