Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001060013.5 | XP_001060013.1 | 7 - 8 | 754 | 103 | |
XM_008766177.1 | XP_008764399.1 | 6 - 7 | 731 | 103 | ||
XM_008766178.1 | XP_008764400.1 | 7 - 8 | 775 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |