Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242963.2 | XP_006243025.1 | 6 | 2562 | 74 | |
XM_236210.8 | XP_236210.3 | 6 | 2452 | 74 | ||
GenBank mRNA | AF091111.1 | - | 1 | 505 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |