Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012501.2 | NP_036633.2 | 1 - 2 | 75 | 78 | |
XM_008766144.1 | XP_008764366.1 | 1 - 2 | 110 | 78 | ||
GenBank mRNA | FQ209522.1 | - | 1 - 2 | 42 | 78 | |
FQ209891.1 | - | 1 - 2 | 44 | 78 | ||
FQ210075.1 | - | 1 - 2 | 75 | 78 | ||
FQ210160.1 | - | 1 - 2 | 44 | 78 | ||
FQ210397.1 | - | 1 - 2 | 42 | 78 | ||
FQ210428.1 | - | 1 - 2 | 46 | 78 | ||
FQ210554.1 | - | 1 - 2 | 78 | |||
FQ210884.1 | - | 1 - 2 | 41 | 78 | ||
FQ211165.1 | - | 1 - 2 | 42 | 78 | ||
FQ211318.1 | - | 1 - 2 | 44 | 78 | ||
FQ211343.1 | - | 1 - 2 | 44 | 78 | ||
FQ211392.1 | - | 1 - 2 | 46 | 78 | ||
FQ211416.1 | - | 1 - 2 | 47 | 78 | ||
FQ211454.1 | - | 1 - 2 | 46 | 78 | ||
FQ218088.1 | - | 1 - 2 | 74 | 78 | ||
FQ218207.1 | - | 1 - 2 | 74 | 78 | ||
FQ218286.1 | - | 1 - 2 | 45 | 78 | ||
FQ219230.1 | - | 1 - 2 | 42 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |