Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139097.3 | NP_620797.1 | 2 - 3 | 435 | 97 | |
XM_006243102.1 | XP_006243164.1 | 2 - 3 | 517 | 97 | ||
XM_006243103.1 | XP_006243165.1 | 2 - 3 | 517 | 97 | ||
XM_008766114.1 | XP_008764336.1 | 2 - 3 | 517 | 97 | ||
GenBank mRNA | AF378093.1 | - | 2 - 3 | 134 | 97 | |
AJ243395.2 | - | 2 - 3 | 420 | 97 | ||
BC070899.1 | - | 2 - 3 | 435 | 7100476 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Rattus norvegicus Probe spans exons |