Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004443.1 | NP_001004443.1 | 10 - 11 | 1181 | 89 | |
XM_006243162.1 | XP_006243224.1 | 11 - 12 | 1051 | 89 | ||
GenBank mRNA | BC082097.1 | - | 10 - 11 | 1181 | 7134440 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |