Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199392.2 | NP_955424.2 | 1 - 2 | 133 | 100 | |
XM_006235578.2 | XP_006235640.1 | 1 - 2 | 167 | 100 | ||
GenBank mRNA | AB096685.1 | - | 1 - 2 | 134 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |