Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106544.1 | NP_001100014.1 | 5 - 6 | 423 | 78 | |
GenBank mRNA | FQ213909.1 | - | 5 - 6 | 401 | 78 | |
FQ214139.1 | - | 5 - 6 | 406 | 78 | ||
FQ216824.1 | - | 5 - 6 | 406 | 78 | ||
FQ219919.1 | - | 5 - 6 | 406 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |