Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017208.2 | NP_058904.2 | 5 - 6 | 603 | 72 | |
GenBank mRNA | BC101906.1 | - | 5 - 6 | 603 | 7933709 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |