Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106539.1 | NP_001100009.1 | 4 - 5 | 927 | 60 | |
XM_006235531.2 | XP_006235593.1 | 4 - 5 | 836 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |