Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106964.1 | NP_001100434.1 | 171 | 67 | ||
GenBank mRNA | BC167084.1 | - | 266 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |