Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Litaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105735.2 | NP_001099205.1 | 1 - 2 | 241 | 89 | |
GenBank mRNA | BC166851.1 | - | 1 - 2 | 241 | 9091522 | 89 |
U53184.1 | - | 278 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |