Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767525.1 | XP_008765747.1 | 5 - 6 | 2478 | 64 | |
XM_008767526.1 | XP_008765748.1 | 5 - 6 | 2421 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |