Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_213210.8 | XP_213210.4 | 9 - 10 | 1083 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |