Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053661.1 | NP_446113.1 | 3 - 4 | 231 | 87 | |
XM_006236610.2 | XP_006236672.1 | 4 - 5 | 795 | 87 | ||
XM_006236612.2 | XP_006236674.1 | 2 - 3 | 196 | 87 | ||
XM_008762969.1 | XP_008761191.1 | 4 - 5 | 695 | 87 | ||
XM_008762970.1 | XP_008761192.1 | 4 - 5 | 670 | 87 | ||
XM_008762971.1 | XP_008761193.1 | 2 - 3 | 848 | 87 | ||
XM_008762972.1 | XP_008761194.1 | 2 - 3 | 696 | 87 | ||
GenBank mRNA | AF022091.1 | - | 1 - 2 | 70 | 87 | |
FM031435.1 | - | 4 - 5 | 249 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |