Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108636.1 | NP_001102106.1 | 3 - 4 | 304 | 148 | |
GenBank mRNA | FQ224589.1 | - | 306 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |