Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029898.1 | NP_001025069.1 | 2 - 3 | 346 | 87 | |
GenBank mRNA | FQ235078.1 | - | 2 - 3 | 364 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |