Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031824.1 | NP_001026994.1 | 1 - 2 | 330 | 96 | |
GenBank mRNA | AB158417.1 | - | 1 - 2 | 330 | 96 | |
AB158418.1 | - | 1 - 2 | 330 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |