Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012735.2 | NP_036867.1 | 15 - 16 | 2414 | 85 | |
XM_006236711.2 | XP_006236773.1 | 14 - 15 | 3483 | 85 | ||
GenBank mRNA | AF027179.1 | - | 15 - 16 | 2218 | 85 | |
M68971.1 | - | 15 - 16 | 2415 | 85 | ||
M68972.1 | - | 7 - 8 | 1016 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |