Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001057850.5 | XP_001057850.2 | 10 - 11 | 5153 | 77 | |
XM_006236792.2 | XP_006236854.1 | 10 - 11 | 4827 | 77 | ||
XM_006236793.2 | XP_006236855.1 | 9 - 10 | 4770 | 77 | ||
XM_006236794.2 | XP_006236856.1 | 9 - 10 | 4196 | 77 | ||
XM_008763094.1 | XP_008761316.1 | 11 - 12 | 4451 | 77 | ||
XM_008763095.1 | XP_008761317.1 | 11 - 12 | 7198 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |