Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236922.2 | XP_006236984.1 | 11 - 12 | 2824 | 69 | |
XM_006236923.2 | XP_006236985.1 | 11 - 12 | 2636 | 69 | ||
XM_006236924.2 | XP_006236986.1 | 10 - 11 | 2683 | 69 | ||
XM_008763139.1 | XP_008761361.1 | 11 - 12 | 2570 | 69 | ||
GenBank mRNA | AB074423.1 | - | 7 - 8 | 915 | 69 | |
AB074424.1 | - | 6 - 7 | 774 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |