Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106606.2 | NP_001100076.2 | 1 - 2 | 62 | 84 | |
GenBank mRNA | FM058785.1 | - | 1 - 2 | 62 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |