Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170546.1 | NP_001164017.1 | 6 - 7 | 1104 | 119 | |
NM_001170547.1 | NP_001164018.1 | 6 - 7 | 1078 | 119 | ||
NM_001170548.1 | NP_001164019.1 | 6 - 7 | 1082 | 119 | ||
XM_006237049.2 | XP_006237111.1 | 6 - 7 | 1453 | 119 | ||
XM_006237050.2 | XP_006237112.1 | 6 - 7 | 1142 | 119 | ||
GenBank mRNA | BC166466.1 | - | 5 - 6 | 1050 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |